Learn More
Medchemexpress LLC Hexahydrofarnesyl acetone | 502-69-2 | 5 MG
SDP
Catalog No. 5000421186
Encompass_Preferred
Hexahydrofarnesyl acetone is an orally active sesquiterpene with antibacterial, anti-inflammatory, and anticancer properties, utilized in pain relief research.
  • Orally active sesquiterpene
  • Demonstrates antibacterial, anti-inflammatory, and anticancer properties
  • Applied in pain relief research
  • Effective against *Staphylococcus aureus* and *Candida albicans* in vitro
  • Exhibits cytotoxicity in various cancer cell lines
  • Inhibits inflammation in animal models
  • Provides analgesic effects in animal models
Catalog No. 50-004-21186 Supplier Medchemexpress LLC Supplier No. HYN30745MG
Hexahydrofarnesyl acetone is an orally active sesquiterpene with antibacterial, anti-inflammatory, and anticancer properties, utilized in pain relief research.
  • Orally active sesquiterpene
  • Demonstrates antibacterial, anti-inflammatory, and anticancer properties
  • Applied in pain relief research
  • Effective against *Staphylococcus aureus* and *Candida albicans* in vitro
  • Exhibits cytotoxicity in various cancer cell lines
  • Inhibits inflammation in animal models
  • Provides analgesic effects in animal models