Learn More
AdipoGen Hexamethonium bromide
SDP
Supplier:  AdipoGen CDXH0455G025
Encompass_Preferred
Chemical. CAS 55-97-0. Formula C12H30Br2N2. MW BD9837. Hexamethonium bromide is a peripherally-acting nondepolarizing neuromuscular blocking agent that acts as a non-competitive antagonist of nicotinic acetylcholine receptors nAChRs. Formulations containing hexamethonium were previously used in the treatment of hypertension. Induces apoptosis and inhibits the stimulatory effect of nicotine on endothelial cell DNA synthesis and proliferation.
Catalog No. 50-256-0685