Learn More
Apexbio Technology LLC Hexamethonium Bromide 55-97-0 1g
SDP
Catalog No. 502240709
Encompass_Preferred
Hexamethonium Bromide (CAS 55-97-0) is a bis-quaternary ammonium compound that acts as a selective antagonist of neuronal nicotinic acetylcholine receptors (nAChRs) located in autonomic ganglia By competitively inhibiting synaptic transmission at these receptors Hexamethonium Bromide effectively blocks ganglionic neurotransmission without significant effects on muscarinic receptors or neuromuscular junctions This property makes it a valuable tool in pharmacological studies investigating autonomic nervous system function and the physiological roles of ganglionic nAChRs in various experimental models
This item is not returnable. View return policy
Catalog No. 50-224-0709 Supplier Apexbio Technology LLC Supplier No. B15921000
Hexamethonium Bromide (CAS 55-97-0) is a bis-quaternary ammonium compound that acts as a selective antagonist of neuronal nicotinic acetylcholine receptors (nAChRs) located in autonomic ganglia By competitively inhibiting synaptic transmission at these receptors Hexamethonium Bromide effectively blocks ganglionic neurotransmission without significant effects on muscarinic receptors or neuromuscular junctions This property makes it a valuable tool in pharmacological studies investigating autonomic nervous system function and the physiological roles of ganglionic nAChRs in various experimental models