Learn More
Medchemexpress LLC HY-B0569 500mg Medchemexpress, Hexamethonium (Bromide) CAS:55-97-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0569500MG
Encompass_Preferred
Medchemexpress, HY-B0569 500mg Hexamethonium (Bromide) CAS:55-97-0 Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) antagonist, with mixed competitive and noncompetitive activity. Hexamethonium Bromide has anti-hypertensive activity. Hexamethonium Bromide attenuates sympathetic activity and blood pressure in spontaneously hypertensive animal models[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2603