Learn More
Medchemexpress LLC Hexamethonium Bromide | 55-97-0 | 99.9% | 100 G
SDP
Catalog No. 5000409464
Encompass_Preferred
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) that exhibits mixed competitive and noncompetitive activity. Known for its anti-hypertensive properties, it can effectively attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models. This product is presented as a high-purity, white to off-white solid.
  • Non-selective ganglionic nAChR antagonist
  • Demonstrates mixed competitive and noncompetitive activity
  • Attenuates sympathetic activity and blood pressure
  • Suitable for research in neurological, cardiovascular, and immune system diseases
  • Available as a white to off-white solid with 99.9% purity
Catalog No. 50-004-09464 Supplier Medchemexpress LLC Supplier No. HYB0569100G
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) that exhibits mixed competitive and noncompetitive activity. Known for its anti-hypertensive properties, it can effectively attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models. This product is presented as a high-purity, white to off-white solid.
  • Non-selective ganglionic nAChR antagonist
  • Demonstrates mixed competitive and noncompetitive activity
  • Attenuates sympathetic activity and blood pressure
  • Suitable for research in neurological, cardiovascular, and immune system diseases
  • Available as a white to off-white solid with 99.9% purity