Learn More
Medchemexpress LLC Hexamethonium Bromide | 55-97-0 | 99.91% | 5 G
SDP
Catalog No. 5000408377
Encompass_Preferred
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor (nAChR) antagonist, exhibiting both competitive and noncompetitive activity. It possesses anti-hypertensive properties and has been shown to reduce sympathetic activity and blood pressure in animal models of spontaneous hypertension.
  • Non-selective ganglionic nicotinic-receptor antagonist
  • Demonstrates mixed competitive and noncompetitive activity
  • Exhibits anti-hypertensive activity
  • Attenuates sympathetic activity
  • Reduces blood pressure in spontaneously hypertensive animal models
Catalog No. 50-004-08377 Supplier Medchemexpress LLC Supplier No. HYB05695G
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor (nAChR) antagonist, exhibiting both competitive and noncompetitive activity. It possesses anti-hypertensive properties and has been shown to reduce sympathetic activity and blood pressure in animal models of spontaneous hypertension.
  • Non-selective ganglionic nicotinic-receptor antagonist
  • Demonstrates mixed competitive and noncompetitive activity
  • Exhibits anti-hypertensive activity
  • Attenuates sympathetic activity
  • Reduces blood pressure in spontaneously hypertensive animal models