Learn More
Medchemexpress LLC Hexamethonium bromide | 55-97-0 | MFCD00008323 | 99.9% | 1 G
SDP
Catalog No. 5000408435
Encompass_Preferred
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor (nAChR) antagonist with mixed competitive and noncompetitive activity. It possesses anti-hypertensive activity and has been shown to attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models.
  • Non-selective ganglionic nicotinic-receptor antagonist (nAChR)
  • Mixed competitive and noncompetitive activity
  • Anti-hypertensive activity
  • Attenuates sympathetic activity and blood pressure in spontaneously hypertensive animal models
Catalog No. 50-004-08435 Supplier Medchemexpress LLC Supplier No. HYB05691G
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor (nAChR) antagonist with mixed competitive and noncompetitive activity. It possesses anti-hypertensive activity and has been shown to attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models.
  • Non-selective ganglionic nicotinic-receptor antagonist (nAChR)
  • Mixed competitive and noncompetitive activity
  • Anti-hypertensive activity
  • Attenuates sympathetic activity and blood pressure in spontaneously hypertensive animal models