Learn More
Medchemexpress LLC Hexamethonium Bromide | 55-97-0 | MFCD00011787 | 99.9% | 10 G
SDP
Catalog No. 5000409022
Encompass_Preferred
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) with mixed competitive and noncompetitive activity. It also exhibits anti-hypertensive activity and can attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models.
  • Target: nAChR (nicotinic-receptor)
  • Research areas: Apoptosis, cardiovascular disease, neurological disease
  • Publications verification: 4 publications verification
Catalog No. 50-004-09022 Supplier Medchemexpress LLC Supplier No. HYB056910G
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) with mixed competitive and noncompetitive activity. It also exhibits anti-hypertensive activity and can attenuate sympathetic activity and blood pressure in spontaneously hypertensive animal models.
  • Target: nAChR (nicotinic-receptor)
  • Research areas: Apoptosis, cardiovascular disease, neurological disease
  • Publications verification: 4 publications verification