Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyl Benzenehexacarboxylate 98.0+%, TCI America™
SDP

Catalog No. B110725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B110725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B110725G Supplier TCI America Supplier No. B110725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6237-59-8
Molecular Formula C18H18O12
Molecular Weight (g/mol) 426.33
MDL Number MFCD00059200
InChI Key BQLICNRRYLYEDI-UHFFFAOYSA-N
Synonym Benzenehexacarboxylic Acid Hexamethyl Ester, Hexamethyl Mellitate, Mellitic Acid Hexamethyl Ester
PubChem CID 138697
IUPAC Name hexamethyl benzene-1,2,3,4,5,6-hexacarboxylate
SMILES COC(=O)C1=C(C(=C(C(=C1C(=O)OC)C(=O)OC)C(=O)OC)C(=O)OC)C(=O)OC

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 25 g
Formula Weight 426.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethyl Benzenehexacarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.