Learn More
Medchemexpress LLC Hexamethylbenzene-d18 | 4342-40-9 | MFCD00144445 | 99.5% | 180.38 g/mol | C12D18 | 100 MG
SDP
Catalog No. 5000301204
Encompass_Preferred
Hexamethylbenzene-d18 is the fully deuterated analogue of hexamethylbenzene (C12D18), supplied as an isotopically enriched reference standard and synthetic intermediate for research. It is characterized by a molecular weight of 180.38 g/mol and is provided as a purified, neat material suitable for analytical, spectroscopic, and mechanistic studies.
  • Fully deuterated (C12D18) for isotope labeling.
  • High isotopic enrichment (~99.3% D) for tracer and mechanistic studies.
  • High chemical purity (HPLC 99.47%) for analytical reliability.
  • Neat, purified material suitable for use as a reference standard or synthetic intermediate.
Catalog No. 50-003-01204 Supplier Medchemexpress LLC Supplier No. HYW038500S100MG
Hexamethylbenzene-d18 is the fully deuterated analogue of hexamethylbenzene (C12D18), supplied as an isotopically enriched reference standard and synthetic intermediate for research. It is characterized by a molecular weight of 180.38 g/mol and is provided as a purified, neat material suitable for analytical, spectroscopic, and mechanistic studies.
  • Fully deuterated (C12D18) for isotope labeling.
  • High isotopic enrichment (~99.3% D) for tracer and mechanistic studies.
  • High chemical purity (HPLC 99.47%) for analytical reliability.
  • Neat, purified material suitable for use as a reference standard or synthetic intermediate.