Learn More
Medchemexpress LLC Hexamethylbenzene-d18 | 4342-40-9 | MFCD00144445 | 99.5% | 180.38 g/mol | C12D18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW038500S
Encompass_Preferred
Hexamethylbenzene-d18 is the deuterium-labeled isotopologue of hexamethylbenzene (C12D18, CAS 4342-40-9) provided for laboratory research use. It appears as a white to off-white solid with high isotopic enrichment and is commonly used as an internal standard, tracer, or reference in mass spectrometry and mechanistic studies. Follow recommended storage conditions for powders or solutions.
  • High isotopic enrichment for accurate mass spectrometry quantitation.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage conditions for long-term use.
  • Suitable as an internal standard, tracer, or reference in research.
  • Available in small pack sizes for laboratory experiments.
Catalog No. 50-003-03961