Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethyldigermanium(IV)

Catalog No. 4476091G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-760-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-760-91G Supplier Sigma Aldrich Supplier No. 4476091G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethyldigermanium(IV)

Specifications

CAS 993-52-2
Melting Point -40°C
Density 1.175 g/mL (at 25°C)
Boiling Point 137°C to 138°C
Molecular Formula C6H18Ge2
Linear Formula (CH3)3 GeGe(CH3)3
Refractive Index n20/D 1.456 (literature)
MDL Number MFCD00047989
Quantity 1 g
Synonym 1,1,1,2,2,2-Hexamethyldigermane; Hexamethyldigermane
Molecular Weight (g/mol) 235.49
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.