Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethyldisilathiane

Catalog No. 2831341G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-313-41G 1 g
28-313-425G 25 g
28-313-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-313-41G Supplier Sigma Aldrich Supplier No. 2831341G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethyldisilathiane may be used in the bis-O-demethylation of dimethoxy aromatic compounds. It may also be used as a sulfur source in the following conversion: Amides and lactams to their corresponding sulfur analogs.Allyl alcohols to diallyl sulfides. Transition metal halides to metal sulfides.Aryl iodides to diaryl sulfides.

Specifications

CAS 3385-94-2
Density 0.846 g/mL (at 25°C)
Boiling Point 164°C
Molecular Formula C6H18SSi2
Refractive Index n20/D 1.4586 (literature)
Linear Formula (CH3)3 SiSSi(CH3)3
Quantity 1 g
MDL Number MFCD00014851
Synonym (TMS)2 S; HMDS; Bis(trimethylsilyl) sulfide; NSC 252160
Molecular Weight (g/mol) 178.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.