Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethyldisilazane

Catalog No. 379212100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-921-2100ML 100 mL
37-921-225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-921-2100ML Supplier Sigma Aldrich Supplier No. 379212100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethyldisilazane (HDMS) is an organoslicon compound.HDMS is a colorless liquid used as a reagent and precursor in organic synthesis and organometallic chemistry.Used in condensation reactions of heterocyclic compounds, such as derivatives of xanthineadhesion promoter for photoresist in photolithographyelectron microscopy, as an alternative to critical point drying during sample preparationpyrolysis-gas chromatography-mass spectrometry, in order to enhance detectability of compounds with polar functional groups.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

Boiling Point 125°C
CAS 999-97-3
Quantity 100 mL
Molecular Formula C6H19NSi2
Linear Formula (CH3)3 SiNHSi(CH3)3
MDL Number MFCD00008259
Synonym 1,1,1,3,3,3-Hexamethyldisilazane; HMDS
Molecular Weight (g/mol) 161.39
Percent Purity 99.9%
Refractive Index n20/D 1.407 (literature)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.