Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethyldisilazane

Catalog No. 869445KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-944-5KGF 5 kg
86-944-500GF 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-944-5KGF Supplier Sigma Aldrich Supplier No. 869445KGF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethyldisilazane (HDMS) is a bulk organo silicon compound.It finds major application as a silanizing agent, silylation with HMDS is carried out in the presence of an acid catalyst.The hydrophobic nature of HDMS coatings on various nanoparticles renders them resistant to water contamination and flocculation's, during synthesis.It has been used as a modifier to control the shape, formation of agglomerates,surface area and pore size of silica particles. precursor for the depositing of SiNx on diamond like carbon films.protective hydrophobic coating for nanoparticles.prices for bulk quantities on request

Specifications

CAS 999-97-3
Boiling Point 125°C
Molecular Formula C6H19NSi2
Refractive Index n20/D 1.407 (literature)
Linear Formula (CH3)3 SiNHSi(CH3)3
Quantity 5 kg
MDL Number MFCD00008259
Synonym Silazane HMN; HMDS
Molecular Weight (g/mol) 161.39
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.