Learn More
Ted Pella Inc Hexamethyldisilazane (HMDS), 999-97-3, MFCD00008259, 250mL
SDP
Catalog No. NC1464993
Encompass
Linear Formula: (CH3)3SiNHSi(CH3)3, Molecular Weight: 161.39, Synonym: 1,1,1,3,3,3-Hexamethyldisilazane, HMDS, BP: 125 °C(lit.). Hexamethyldisilazane (HDMS) is an organoslicon compound.HDMS is a colorless liquid used as a reagent and precursor in organic synthesis and organometallic chemistry.
Catalog No. NC1464993 Supplier Ted Pella Inc Supplier No. 18605
Linear Formula: (CH3)3SiNHSi(CH3)3, Molecular Weight: 161.39, Synonym: 1,1,1,3,3,3-Hexamethyldisilazane, HMDS, BP: 125 °C(lit.). Hexamethyldisilazane (HDMS) is an organoslicon compound.HDMS is a colorless liquid used as a reagent and precursor in organic synthesis and organometallic chemistry.