Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethyldisiloxane

Catalog No. 46930050ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-930-050ML 50 mL
46-930-0250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-930-050ML Supplier Sigma Aldrich Supplier No. 46930050ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethyldisiloxane

Specifications

CAS 107-46-0
Melting Point -59°C
Density 0.764 g/mL (at 20°C)
Boiling Point 101°C
Molecular Formula C6H18OSi2
Refractive Index n20/D 1.377 (literature)
Linear Formula (CH3)3 SiOSi(CH3)3
Quantity 50 mL
MDL Number MFCD00008265
Synonym Poly(dimethylsiloxane); HMDSO
Molecular Weight (g/mol) 162.38
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.