Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisiloxane 98.0+%, TCI America™
SDP

Catalog No. H0091100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0091100ML 100 mL
H009125ML 25 mL
H0091500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0091100ML Supplier TCI America Supplier No. H0091100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-46-0
Molecular Formula C6H18OSi2
Molecular Weight (g/mol) 162.379
MDL Number MFCD00008265
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 100°C
Quantity 100 mL
UN Number 1993
Formula Weight 162.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexamethyldisiloxane
EINECSNumber (2)-2956
RTECSNumber JM9237000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.