Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylditin, 97%

Catalog No. AAA1763814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1763814 25 g
AAA1763803 1 g
AAA1763806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1763814 Supplier Thermo Scientific Chemicals Supplier No. A1763814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 661-69-8 | C6H18Sn2 | 327.63 g/mol

It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents, heat and air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-69-8
Molecular Formula C6H18Sn2
Molecular Weight (g/mol) 327.63
MDL Number MFCD00008277
InChI Key CCRMAATUKBYMPA-UHFFFAOYSA-N
Synonym hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci
PubChem CID 6327618
SMILES C[Sn](C)(C)[Sn](C)(C)C

Specifications

Melting Point 23°C
Density 1.58
Boiling Point 85°C (45 mmHg)
Flash Point 61°C (142°F)
Odor Characteristic
Refractive Index 1.541
Linear Formula (CH3)3SnSn(CH3)3
Quantity 25 g
UN Number UN2788
Beilstein 3600327
Sensitivity Air sensitive
Solubility Information Not miscible or difficult to mix in water.
IUPAC Name trimethyltin
Formula Weight 327.59
Percent Purity 97%
Chemical Name or Material Hexamethylditin
Show More Show Less
Hazard Category H227-H300+H310+H330-H335-H336-H373
Hazard Statement GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 211-549-2
RTECSNumber WH8280000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.