Learn More
Cayman Chemical Hexamethylene bisacetamide, Purity: ≥95%, CAS: 3073-59-4, 50 g
SDP
Catalog No. 502988089
Encompass_Preferred
A tumor cell-differentiating agent; induces complete differentiation of 754A murine erythroleukemia cells at 5 mM; induces latent HIV-1 viral production in chronically HIV-1-infected U1 cells; implantation of HT-29 cells cultured with hexamethylene bisacetamide for seven, but not 28, days reduces tumorigenesis of those cells in mice
Catalog No. 50-298-8089 Supplier Cayman Chemical Supplier No. 3085950 G
A tumor cell-differentiating agent; induces complete differentiation of 754A murine erythroleukemia cells at 5 mM; induces latent HIV-1 viral production in chronically HIV-1-infected U1 cells; implantation of HT-29 cells cultured with hexamethylene bisacetamide for seven, but not 28, days reduces tumorigenesis of those cells in mice