Learn More
Lkt Laboratories Hexamethylene Bisacetamide, ≥99%, 25G
SDP
Catalog No. NC3371263
Encompass_Preferred
Hexamethylene bisacetamide (HMBA) activates hexamethylene bisacetamide-inducible protein 1 (HEXIM1), inhibiting transcription elongation factor b (P-TEFb), a regulator of RNA polymerase II and mRNA synthesis. HMBA exhibits anticancer activity and is used to induce cellular differentiation and apoptosis in cancer cells. HMBA also inhibits activation of NF-κB, Akt, and ERK. CAS: 3073-59-4. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC3371263 Supplier Lkt Laboratories Supplier No. H1892
Hexamethylene bisacetamide (HMBA) activates hexamethylene bisacetamide-inducible protein 1 (HEXIM1), inhibiting transcription elongation factor b (P-TEFb), a regulator of RNA polymerase II and mRNA synthesis. HMBA exhibits anticancer activity and is used to induce cellular differentiation and apoptosis in cancer cells. HMBA also inhibits activation of NF-κB, Akt, and ERK. CAS: 3073-59-4. Additional sizes available upon request. Ships from Saint Paul, MN.