Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexamethylene bisacetamide | 3073-59-4 | MFCD00008684 | 99.6% | 200.28 | C10H20N2O2 | 100MG
SDP

Catalog No. 5000238550
Encompass_Preferred

Hexamethylene bisacetamide (HMBA) is a small-molecule differentiation inducer used in cell biology and cancer research to promote terminal differentiation and modulate signaling pathways. It is provided as a high-purity solid suitable for in vitro experiments and biochemical studies, with specified solubility and storage conditions for short- and long-term use.

  • High purity (99.6%).
  • Molecular weight 200.28.
  • Chemical formula C10H20N2O2.
  • Soluble in water (100 mg/mL) and DMSO (25 mg/mL) with ultrasonic assistance.
  • Appearance: white to off-white solid, supplied in 100 mg packaging.
  • Storage: long-term 4°C protected from light; in solution -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-38550 Supplier Medchemexpress LLC Supplier No. HY124284100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexamethylene bisacetamide (HMBA) is a small-molecule differentiation inducer used in cell biology and cancer research to promote terminal differentiation and modulate signaling pathways. It is provided as a high-purity solid suitable for in vitro experiments and biochemical studies, with specified solubility and storage conditions for short- and long-term use.

  • High purity (99.6%).
  • Molecular weight 200.28.
  • Chemical formula C10H20N2O2.
  • Soluble in water (100 mg/mL) and DMSO (25 mg/mL) with ultrasonic assistance.
  • Appearance: white to off-white solid, supplied in 100 mg packaging.
  • Storage: long-term 4°C protected from light; in solution -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.