Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexamethylene bisacetamide | 3073-59-4 | MFCD00008684 | 99.6% | 200.28 g/mol | C10H20N2O2 | 10MM 1ML
SDP

Catalog No. 5000239196
Encompass_Preferred

Hexamethylene bisacetamide (HMBA) is a small-molecule cell differentiation inducer used in biochemical and cellular research. It is available as a ready-to-use 10 mM solution in 1 mL DMSO or as solid quantities, with molecular formula C10H20N2O2 and molecular weight 200.28 g/mol.

  • Differentiation inducer for cell biology studies.
  • Supplied as 10 mM in 1 mL DMSO or as solid quantities.
  • High purity suitable for research (≈99.6%).
  • Soluble in DMSO for easy formulation of working solutions.
  • Store solids protected from light at 4°C; in solution store at -80°C for up to 6 months.
  • Molecular weight 200.28 g/mol; CAS 3073-59-4.

Catalog No. 50-002-39196 Supplier Medchemexpress LLC Supplier No. HY12428410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexamethylene bisacetamide (HMBA) is a small-molecule cell differentiation inducer used in biochemical and cellular research. It is available as a ready-to-use 10 mM solution in 1 mL DMSO or as solid quantities, with molecular formula C10H20N2O2 and molecular weight 200.28 g/mol.

  • Differentiation inducer for cell biology studies.
  • Supplied as 10 mM in 1 mL DMSO or as solid quantities.
  • High purity suitable for research (≈99.6%).
  • Soluble in DMSO for easy formulation of working solutions.
  • Store solids protected from light at 4°C; in solution store at -80°C for up to 6 months.
  • Molecular weight 200.28 g/mol; CAS 3073-59-4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.