Learn More
Medchemexpress LLC Hexamethylene bisacetamide | 3073-59-4 | MFCD00008684 | 99.6% | 200.28 g·mol⁻¹ | C10H20N2O2 | 50MG
SDP
Catalog No. 5000235369
Encompass_Preferred
Hexamethylene bisacetamide (HMBA) is a small-molecule differentiation inducer and bromodomain inhibitor used as a research reagent to promote cell differentiation, inhibit proliferation, and investigate apoptosis-related signaling pathways in cell biology and cancer research. It is provided for laboratory use in small pack sizes suitable for in vitro and analytical applications.
  • Research-grade differentiation inducer for cell biology assays.
  • Reported to cross the blood-brain barrier in preclinical studies.
  • Induces apoptosis via Notch1, Bcl-2, and p53 signaling pathways.
  • High purity for analytical and in vitro applications (≈99.6%).
  • Chemical formula C10H20N2O2 and molecular weight 200.28 g·mol⁻¹.
  • Available in small pack sizes suitable for research workflows.
Catalog No. 50-002-35369 Supplier Medchemexpress LLC Supplier No. HY12428450MG
Hexamethylene bisacetamide (HMBA) is a small-molecule differentiation inducer and bromodomain inhibitor used as a research reagent to promote cell differentiation, inhibit proliferation, and investigate apoptosis-related signaling pathways in cell biology and cancer research. It is provided for laboratory use in small pack sizes suitable for in vitro and analytical applications.
  • Research-grade differentiation inducer for cell biology assays.
  • Reported to cross the blood-brain barrier in preclinical studies.
  • Induces apoptosis via Notch1, Bcl-2, and p53 signaling pathways.
  • High purity for analytical and in vitro applications (≈99.6%).
  • Chemical formula C10H20N2O2 and molecular weight 200.28 g·mol⁻¹.
  • Available in small pack sizes suitable for research workflows.