Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethylenetetramine

Catalog No. H113004KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
4 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H113004KG 4 kg
H11300500G 500 g
H1130025G 25 g
H113001KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. H113004KG Supplier Sigma Aldrich Supplier No. H113004KG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethylenetetramine

Specifications

CAS 680-31-9
Melting Point 7°C
Density 1.03 g/mL at 25°C
Boiling Point 230°C to 232°C (740 mmHg, lit.)
Molecular Formula C6H18N3OP
Linear Formula [(CH3)2N]3PO
Quantity 4 kg
MDL Number MFCD00008303
Synonym 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane; Hexamine; Methenamine; Urotropine
Molecular Weight (g/mol) 179.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.