Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Hexamethylenetetramine 50 GR
SDP

Catalog No. 5026499
Encompass_Preferred

(Urotropine; Methenamine)
CAS #100-97-0 C₆N₄ F.W. 140.19
Assay: 99.0-100.5%

Hexamethylenetetramine (Urotropine; Methenamine) is used in silver stains. Hexamethylenetetramine is also available in 25 ml bottles to avoid its classification as a hazardous chemical for shipping purposes.

Catalog No. 50-264-99 Supplier Electron Microscopy Sciences Supplier No. 16710
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Urotropine; Methenamine)
CAS #100-97-0 C₆N₄ F.W. 140.19
Assay: 99.0-100.5%

Hexamethylenetetramine (Urotropine; Methenamine) is used in silver stains. Hexamethylenetetramine is also available in 25 ml bottles to avoid its classification as a hazardous chemical for shipping purposes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.