Learn More
Hach Company Hexamethylenetetramine Buffer Reagent Powder Pillows, pk/100
Catalog No. 501996320 Shop All Hach Company Products
Encompass
For aluminum determination by the Eriochrome Cyanine R method (Hach method 8326). Range: 0.002 - 0.250 mg/L Al Sample Size: 20 mL (split) Pack of 100 powder pillows.
Catalog No. 50-199-6320 Supplier Hach Company Supplier No. 2603999
For aluminum determination by the Eriochrome Cyanine R method (Hach method 8326). Range: 0.002 - 0.250 mg/L Al Sample Size: 20 mL (split) Pack of 100 powder pillows.