Learn More
Hach Company Hexamethylenetetramine Buffer Reagent Powder Pillows, pk/50
Catalog No. 501994623 Shop All Hach Company Products
Encompass
For aluminum determination by the Eriochrome Cyanine R method (Hach method 8326). Sample Size: 25 mL. Pack of 50 powder pillows.
Catalog No. 50-199-4623 Supplier Hach Company Supplier No. 187866
For aluminum determination by the Eriochrome Cyanine R method (Hach method 8326). Sample Size: 25 mL. Pack of 50 powder pillows.