Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethylphosphoramide

Catalog No. H11602100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11602100G 100 g
H1160225KG 2.5 kg
H11602500G 500 g
H116025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. H11602100G Supplier Sigma Aldrich Supplier No. H11602100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethylphosphoramide

Specifications

CAS 7389-87-9
Melting Point 207°C
Molecular Formula C7H11N3O2 -+ 2HCl
Refractive Index n20/D 1.459
Linear Formula C7H11N3O2 -+ 2HCl
MDL Number MFCD00012701
Quantity 100 g
Synonym HMPA; Hexamethylphosphoric acid triamide; Tris(dimethylamino)phosphine oxide
Molecular Weight (g/mol) 242.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.