Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexanal

Catalog No. 115606250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
100 mL
250 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-560-6250ML 250 mL
11-560-62ML 2 mL
11-560-61L 1 L
11-560-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-560-6250ML Supplier Sigma Aldrich Supplier No. 115606250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hexanal is present in lactoperoxidase-bleached wpc80 (whey protein concentrate with 80% protein).hexanal has been used in the quantification of carbonyls that contribute to the complex flavor of olive oil
  • It can also be used for wittig and aldol reactions

Specifications

CAS 66-25-1
Density 0.815 g/mL at 25°C (literature); 0.834 g/mL (at 25°C)
Boiling Point 130°C to 131°C (lit.)
Molecular Formula C6H12O
Refractive Index n20/D 1.4035 (literature)
Linear Formula CH3(CH2)4 CHO
MDL Number MFCD00007027
Quantity 250 mL
Synonym Aldehyde C6; Caproaldehyde; Hexyl aldehyde
Molecular Weight (g/mol) 100.16
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.