Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanal, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019884 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9884 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9884 Supplier MilliporeSigma Supelco Supplier No. 181091ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexanal is an oxidation product of linoleic acid and is a dominant volatile aldehyde. It could work as an indicator of lipid oxidation for cooked meat and meat flavor deterioration.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 66-25-1
Vapor Pressure 10 mmHg (20°C)
Vapor Density >1 (vs. air)
Density 0.815 g/mL (at 25°C)
Flash Point 25°C
Physical Form Neat
Linear Formula CH3(CH2)4 CHO
MDL Number MFCD00007027
Boiling Point 130°C to 131°C
Synonym Aldehyde C6; Caproaldehyde; Hexyl aldehyde
Type Analytical Standard
Quantity 1 mL
Formula Weight 100.16
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.