Learn More
AccuStandard, Inc HEXANAL-DNPH 1000UG/ML 1ML
SDP
Supplier:  AccuStandard, Inc M554DNPH08
Encompass
Hexanal-DNPH. 1 mL. 1000 �g/mL in MeOH:AcCN (95:5) (as deriv.). 1527-97-5
Catalog No. NC1047358