Learn More
Medchemexpress LLC Hexanamide, 2-(acetylamino)-6-amino-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-, (2S)- | 156661-42-6 | 98.9% | 345.39 g/mol | C18H23N3O4 | 5 MG
SDP
Catalog No. 5000262958
Encompass_Preferred
Ac-Lys-AMC (Hexanamide), also termed MAL, is a fluorogenic substrate used to measure histone deacetylase (HDAC) activity. After HDAC-catalyzed deacetylation and subsequent enzymatic cleavage, the 7-amido-4-methylcoumarin (AMC) moiety is released, producing a fluorescent signal for quantitative biochemical assays and inhibitor screening. Supplied as a solid for research use only.
  • Fluorogenic substrate for HDAC activity assays.
  • Enables sensitive fluorescence detection via AMC release.
  • High purity (HPLC) for reliable assay results.
  • Solid, white to off-white physical form for easy storage and handling.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-62958 Supplier Medchemexpress LLC Supplier No. HY1289195MG
Ac-Lys-AMC (Hexanamide), also termed MAL, is a fluorogenic substrate used to measure histone deacetylase (HDAC) activity. After HDAC-catalyzed deacetylation and subsequent enzymatic cleavage, the 7-amido-4-methylcoumarin (AMC) moiety is released, producing a fluorescent signal for quantitative biochemical assays and inhibitor screening. Supplied as a solid for research use only.
  • Fluorogenic substrate for HDAC activity assays.
  • Enables sensitive fluorescence detection via AMC release.
  • High purity (HPLC) for reliable assay results.
  • Solid, white to off-white physical form for easy storage and handling.
  • Intended for research use only; not for human or clinical use.