Learn More
Medchemexpress LLC Hexanamide, N-[(1S,2R,3E)-1-[(β-D-glucopyranosyloxy)methyl]-2-hydroxy-3-heptadecen-1-yl]-6-[(7-nitro | 94885-03-7 | 98.0% | 737.88 g·mol⁻¹ | C36H59N5O11 | 100 UG
SDP
Catalog No. 5000285210
Encompass_Preferred
C6 NBD Glucosylceramide is a fluorescently labeled glucosylceramide derivative used to investigate sphingolipid metabolism and intracellular trafficking. The NBD fluorophore on a C6 acyl chain provides fluorescence properties suitable for imaging and plate-based assays, and the material is supplied at high purity for biochemical experiments.
  • Fluorescent label suitable for microscopy and plate assays.
  • Excitation 466 nm and emission 535 nm for standard fluorescence setups.
  • High purity suitable for biochemical and enzymatic assays (98.0%).
  • Useful for studying glucosylceramide metabolism and internalization.
  • Available in small-scale vials ideal for research applications (100 μg, 500 μg).
Catalog No. 50-002-85210 Supplier Medchemexpress LLC Supplier No. HYW356117100UG
C6 NBD Glucosylceramide is a fluorescently labeled glucosylceramide derivative used to investigate sphingolipid metabolism and intracellular trafficking. The NBD fluorophore on a C6 acyl chain provides fluorescence properties suitable for imaging and plate-based assays, and the material is supplied at high purity for biochemical experiments.
  • Fluorescent label suitable for microscopy and plate assays.
  • Excitation 466 nm and emission 535 nm for standard fluorescence setups.
  • High purity suitable for biochemical and enzymatic assays (98.0%).
  • Useful for studying glucosylceramide metabolism and internalization.
  • Available in small-scale vials ideal for research applications (100 μg, 500 μg).