Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc HEXANE -0.6 EC 200L
SDP

Catalog No. 50326240 Shop All Jade Scientific, Inc Products
Encompass_Preferred

Hexane -0.6 Ec 200l. 110-54-3 MFCD00009520

Catalog No. 50-326-240 Supplier Jade Scientific, Inc Supplier No. BJCS219200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexane -0.6 Ec 200l. 110-54-3 MFCD00009520

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.