Learn More
Medchemexpress LLC Hexane, 1-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]-6-chloro- | 1625717-44-3 | MFCD34471373 | 95.3% | 293.79 | C12H24ClN3O3 | 5 MG
SDP
Catalog No. 5000222859
Encompass_Preferred
Azido-PEG3-C6-Cl is a PEG-based linker bearing an azide group used for click chemistry and PROTAC synthesis. It facilitates conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), providing a versatile building block for bioconjugation and linker design.
  • Contains an azide functional group for efficient CuAAC reactions.
  • Compatible with strain-promoted azide-alkyne cycloaddition for catalyst-free conjugation.
  • PEG3 spacer improves solubility and provides linker flexibility.
  • Terminal chlorohexyl enables further chemical modification.
  • High purity and characterized molecular data support synthetic use.
Catalog No. 50-002-22859 Supplier Medchemexpress LLC Supplier No. HY1384665MG
Azido-PEG3-C6-Cl is a PEG-based linker bearing an azide group used for click chemistry and PROTAC synthesis. It facilitates conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), providing a versatile building block for bioconjugation and linker design.
  • Contains an azide functional group for efficient CuAAC reactions.
  • Compatible with strain-promoted azide-alkyne cycloaddition for catalyst-free conjugation.
  • PEG3 spacer improves solubility and provides linker flexibility.
  • Terminal chlorohexyl enables further chemical modification.
  • High purity and characterized molecular data support synthetic use.