Learn More
Medchemexpress LLC Hexane-1,6-diol | 629-11-8 | 99.86% | 118.17 | 1000 G
SDP
Catalog No. 5000451872
Encompass_Preferred
Hexane-1,6-diol is a 6-carbon straight-chain diol with hydroxyl groups at both ends. It can be used as an additive for drilling fluids and as a substrate in life science-related research. Its monoesters and diesters have been used as substrates for in vitro digestion studies by pancreatic lipase, undergoing hydrolysis and transesterification decomposition pathways. Hexane-1,6-diol can also be catalytically lactonized by alcohol dehydrogenases like AaSDR-1 to produce ε-caprolactone, with its conversion influenced by buffers.
  • 6-carbon straight-chain diol
  • Hydroxyl groups at both ends
  • Can be used as an additive for drilling fluids
  • Substrate in life science-related research
  • Used in in vitro digestion studies by pancreatic lipase
  • Can be catalytically lactonized to produce ε-caprolactone
Catalog No. 50-004-51872 Supplier Medchemexpress LLC Supplier No. HYW0105051000G
Hexane-1,6-diol is a 6-carbon straight-chain diol with hydroxyl groups at both ends. It can be used as an additive for drilling fluids and as a substrate in life science-related research. Its monoesters and diesters have been used as substrates for in vitro digestion studies by pancreatic lipase, undergoing hydrolysis and transesterification decomposition pathways. Hexane-1,6-diol can also be catalytically lactonized by alcohol dehydrogenases like AaSDR-1 to produce ε-caprolactone, with its conversion influenced by buffers.
  • 6-carbon straight-chain diol
  • Hydroxyl groups at both ends
  • Can be used as an additive for drilling fluids
  • Substrate in life science-related research
  • Used in in vitro digestion studies by pancreatic lipase
  • Can be catalytically lactonized to produce ε-caprolactone