Learn More
Medchemexpress LLC Hexane-1,6-diol | 629-11-8 | 99.9% | 118.17 | 1 ML
SDP
Catalog No. 5000366365
Encompass_Preferred
Hexane-1,6-diol is a 6-carbon straight-chain diol with hydroxyl groups at both ends. It can be used as an additive for drilling fluids and as a substrate in life science-related research. For research use only; not sold to patients.
  • Monoesters and diesters act as substrates for in vitro digestion studies by pancreatic lipase.
  • Can be catalytically lactonized by alcohol dehydrogenases like AaSDR-1 to produce ε-caprolactone.
Catalog No. 50-003-66365 Supplier Medchemexpress LLC Supplier No. HYW01050510MM/1ML
Hexane-1,6-diol is a 6-carbon straight-chain diol with hydroxyl groups at both ends. It can be used as an additive for drilling fluids and as a substrate in life science-related research. For research use only; not sold to patients.
  • Monoesters and diesters act as substrates for in vitro digestion studies by pancreatic lipase.
  • Can be catalytically lactonized by alcohol dehydrogenases like AaSDR-1 to produce ε-caprolactone.