Learn More
Medchemexpress LLC Hexane-1,6-diyldiphosphonic acid | 4721-22-6 | MFCD01630742 | 98% | C6H16O6P2 | 100 MG
SDP
Catalog No. 5000357336
Encompass_Preferred
Hexane-1,6-diyldiphosphonic acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • Alkyl chain-based PROTAC linker
  • Can be utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-57336 Supplier Medchemexpress LLC Supplier No. HY140326100MG
Hexane-1,6-diyldiphosphonic acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • Alkyl chain-based PROTAC linker
  • Can be utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins