Learn More
Hexane CHROMASOLV™, for analysis of dioxins, furans and PCB, ≥95%, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- High purity chromatography solvents
- Suitability tested for specified use from HPLC up to UHPLC-MS applications.
Specifications
Specifications
| Density | 0.659g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 0.077 |
| Flash Point | −26°C (−14.8°F) |
| Ignition Point | 453°F |
| Linear Formula | CH3(CH2)4CH3 |
| Vapor Pressure | 256 mmHg (37.7°C), 5.2 psi (37.7°C), 132 mmHg (20°C) |
| UN Number | UN1208 |
| Beilstein | 1730733 |
| Formula Weight | 86.18g/mol |
| Show More |
Safety and Handling
| Hazard Statement | P201-P210-P273-P301 + P310-P308 + P313-P331 |
| RTECSNumber | MN9275000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.