Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane), Solstice

Catalog No. 6001078 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-010-78 4 x 4 L Glass Bottle
60-010-79 4 x 4 L Poly Bottle
60-010-76 19 L Metal Can
60-010-77 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-010-78 Supplier Solstice Supplier No. LP2114
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Min % >60.0000%
CAS Max % >60.00%, <40.0000%
Density 0.659 - 0.673 g/cm3
Quantity 4 x 4 L
Flash Point −26°C
Packaging Glass Bottle
Water 0.01% max.
UN Number UN1208
Beilstein 1730733
Boiling Point 68.7°C
Solubility Information 1.4% in Water at 20°C
Formula Weight 86.18
Percent Purity >60%
Grade ACS
Assay by GC Analysis n-Hexane
Chemical Name or Material Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.