Learn More
Supply Solutions HEXANE, ANHYDROUS, 9
Supplier:  Supply Solutions 2960902L
Encompass_Preferred
Hexane, anhydrous, 95%110-54-3MFCD00009520Linear Formula CH3(CH2)4CH3, Molecular Weight 86.18, Synonym: n-Hexane
Catalog No. NC1684726