Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, For pesticide residue analysis, Solstice

Catalog No. 60046345 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-345 4 x 2.5 L
60-010-55 1 L
60-010-57 4 x 4 L
60-010-56 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-345 Supplier Solstice Supplier No. 344844X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.659 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point −26°C
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
UN Number UN1208
Beilstein 1730733
Boiling Point 66°C to 69°C
Formula Weight 86.18
Percent Purity >95%
Grade Pesticide
Chemical Name or Material Hexane, For pesticide residue analysis
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes
EINECSNumber 203-777-6
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.