Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, CHROMASOLV Plus, for HPLC, mixture of isomers,≥98.5%, Solstice

Catalog No. 60090310 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-310 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-310 Supplier Solstice Supplier No. 650544200L
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 86.18

Specifications

CAS Max % <40.00
CAS 110-54-3
CAS Min % >60.00
Density 0.659 - 0.673 g/cm3 (20°C)
Quantity 200 L
Flash Point -26°C
Packaging Returnable Steel Drum
Physical Form Liquid
Boiling Point 68.7°C
Grade HPLC
Hazard Category Danger
Hazard Statement H225+H304+H315+H336+H361+H373
Precautionary Statement P280+P271+P264+P260+P243+P242+P241+P240+P233+P210+P202+P201
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.