Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Solstice

Catalog No. 60090303 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
Packaging:
Returnable Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-303 56 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-303 Supplier Solstice Supplier No. CS212AH56
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9% (>60% n-Hexane)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Max % <40.00%
CAS Min % >60.00%
Quantity 56 L
Flash Point -26°C
Packaging Returnable Steel Drum
Physical Form Liquid, clear
Boiling Point 68.7°C
Grade Performance HPLC solvents
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.