Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Solstice

Catalog No. 60090074 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-074 56 L
60-090-073 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-074 Supplier Solstice Supplier No. CS21356
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For GC, for pesticide residue analysis, non-spectrophotometric grade, >95% n-Hexane, >99.9% (>95% n-Hexane)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Max % <40.00%
CAS Min % >60.00%
Quantity 56 L
Flash Point -26°C
Packaging Returnable Steel Drum
Physical Form Liquid, clear
Boiling Point 68.7°C
Grade Multipurpose solvents
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.