Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Solstice

Catalog No. 60090302 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-302 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-302 Supplier Solstice Supplier No. 2154X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Capillary GC, GC-FID, GC-MS, >95% n-Hexane, >99.9%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

PubChem CID 8058
ChEBI CHEBI:29021

Specifications

CAS Max % <40.00%
CAS 110-54-3
CAS Min % >60.00%
Quantity 4 x 4 L
Flash Point -26°C
Packaging Glass Bottle
Physical Form Liquid, clear
Boiling Point 68.7°C
Grade Multipurpose solvents
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.