Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Laboratory Reagent, ≥95%, Solstice

Catalog No. p-7120459 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
4 x 2.5 L
4 x 5 L
6 x 1 L
190 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Stainless Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-160 1 L Glass Bottle
60-045-161 2.5 L Glass Bottle
60-045-168 6 x 1 L Glass Bottle
60-045-164 4 x 2.5 L Glass Bottle
60-045-159 190 L Metal Drum
60-045-165 4 x 5 L Aluminum Bottle
60-045-162 20 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60-045-160 Supplier Solstice Supplier No. 2087521L
Only null left
Add to Cart
Add to Cart

Laboratory Reagent,≥95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.659g/mL at 25°C
Quantity 1 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.375(lit.)
Vapor Pressure 132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C)
Linear Formula CH3(CH2)4CH3
UN Number UN1208
Beilstein 1730733
Boiling Point 69°C
Formula Weight 86.18g/mol
Percent Purity ≥95%
Grade Laboratory Reagent
Chemical Name or Material Hexane
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H361f-H373-H411
DOTInformation Class 3 - PG 2 - Hexanes
EINECSNumber 203-777-6
RTECSNumber MN9275000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.