Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Mixture of isomers for HPLC, ≥98.5%, MilliporeSigma™ Supelco™

Catalog No. 111005334 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-5334 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5334 Supplier MilliporeSigma Supelco Supplier No. 293253200LP1SBYE
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Contains thiophene

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -95°C
Color APHA: ≤10
Density 0.672 g/mL (at 25°C)
Quantity 200 L
Packaging Glass Bottle
Refractive Index n20/D 1.379
Linear Formula C6H14
MDL Number MFCD02179311
Synonym Hexanes; Isohexane
Boiling Point 68°C to 70°C
Formula Weight 86.18
Percent Purity ≥98.5%
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.