Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexane mixture of isomers
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1789184L
Encompass
The product is a mixture of hexane isomers and methylcyclopentane. Hexane (mixture of isomers) can be isolated from petroleum by distillation or by cracking. They form major constituents of petroleum ether. It is a non-renewable organic solvent mainly used for the disSOLUTIONon and extraction of oils and fats. Separation of the isomeric forms of hexane on the basis of degree of branching using fe2(BDP)3 (BDP2- 1 4-benzenedipyrazolate) has been reported.
Catalog No. 50-289-7549